missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Novex™ Zymogram Plus (Gelatin) Protein Gels, 10%, 1.0 mm

Product Code. p-7237815
Click to view available options
No. of Wells:
10-well
12-well
15-well
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15605608

Brand: Novex™ ZY00100BOX

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Novex 10% Zymogram Plus (Gelatin) gels are useful for the detection and characterization of proteases that use gelatin as a substrate. The proteases are run under denaturing conditions and visualized as clear bands against a dark background using a simple renaturing, developing, and staining protocol.

Novex 10% Zymogram Plus (Gelatin) gels are useful for the detection and characterization of proteases that use gelatin as a substrate. The proteases are run under denaturing conditions and visualized as clear bands against a dark background using a simple renaturing, developing, and staining protocol. Zymogram gels are commonly used to detect matrix metalloproteinases. Novex 10% Zymogram Plus gels are highly sensitive, detecting as little as 5 x 10-6 units of collagenase.

Formulation
Novex Zymogram Plus gels are Tris-glycine gels with 0.1% gelatin incorporated into the gel. They are made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, highly purified water, and gelatin as the substrate. All have a 4% stacking gel.

Choose the right gel format for your experiments
Novex 10% Zymogram Plus gels come in a fixed concentration of 10% gelatin and three different well formats, including 10-, 12- and 15-well.

Recommended buffers
Use Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC2675) with these gels. Novex Zymogram Renaturing Buffer (LC2670) and Novex Zymogram Developing Buffer (LC2671) are also recommended.

Recommended Storage

Store in refrigerator (2-8°C). Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mini Gel Tank, XCell SureLock™ Mini-Cell
Gel Size Mini
Product Line Novex
Gel Percentage 0.1
Gel Type Zymogram Gel
Mode of Separation Molecular Weight
No. of Wells 10-well
Quantity 10 gels (1 box)
Sample Loading Volume Up to 25 μL
Separation Range 30 to 150 kDa (Tris-Glycine Buffer)
Shipping Condition Wet Ice
Temperature (Metric) Storage Store at 2°C to 8°C. Do not freeze.
Thickness (Metric) 1.0 mm
Width (Metric) 8 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.