missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 4X Bolt™ LDS Sample Buffer

Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats

Brand:  Invitrogen™ B0007

14.59 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 13276499

  • 20.55 € / 10mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: lithium dodecyl sulfate (LDS) at pH 8.4.
This item is not returnable. View return policy

Description

Description

Novex™ Bolt™ buffers and sample reducing agents are optimized for use with Bolt Bis-Tris Plus gels, and are available in a variety of formats.

About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.

Bolt™ gel run times may vary depending upon the type of running buffer used.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

4X
Bis-Tris Plus
Bolt™ Bis-Tris Plus Gels
10 mL
Sample Loading Buffers
Bolt Mini Gel Tank
LDS Sample Buffer
Bolt™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3051981 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberB0007,B0007_LITL18 PROMO
Lot Number2463557 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberB0007,B0007_LITL18 PROMO
Lot Number2491553 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberB0007,B0007_LITL18 PROMO
Lot Number2544541 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberB0007,B0007_LITL18 PROMO
Lot Number2552746 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberB0007,B0007_LITL18 PROMO
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ 4X Bolt™ LDS Sample Buffer > 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.