missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human beta Amyloid (1-40) PTD Recombinant Protein
Click to view available options
Quantity:
1 mg
Packungsgröße:
Each
Beschreibung
Human beta Amyloid (1-40) PTD Recombinant Protein for Bioactivity
Beta-amyloid is a 40-43 amino acid peptide cleaved from amyloid precursor protein, and a putative gamma-secretase. Amyloid beta (1-40) together with Amyloid beta (1-42) are two major C-terminal variants of the amyloid beta protein, constituting the majority of amyloid beta proteins. Both the amyloid beta (40) and amyloid beta (42) alloforms are major components of the extracellular plaques found in Alzheimer's disease brain tissue.
Spezifikation
Spezifikation
| Accession Number | P05067 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 351 |
| Molecular Weight (g/mol) | 4.3 kDa |
| Name | Human beta Amyloid (1-40) PTD |
| Purification Method | Purified |
| Quantity | 1 mg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur