missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Novex™ AP Chromogenic Substrate (BCIP/ NBT)

Novex™ AP Chromogenic Substrate (BCIP/ NBT)

Brand:  Novex™ WP20001


Product Code. 10781384

  • 205.00 € / 250mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Novex™ AP Chromogenic Substrate is a ready to use single component reagent for sensitive immunodetection of alkaline phosphatase (AP) enzyme on western blot and dot blot membranes. Compatable for use with Western Breeze™ protocols, this ready-to-use solution of 5-bromo-4-chloro-3-indolyl-1-phosphate (BCIP) and nitroblue tetrazolium (NBT) will forms a black-purple precipitate upon reaction with alkaline phosphatase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single bottle labeled: Novex™ AP Chromogenic Substrate (BCIP⁄NBT). Store at 4°C.
Liquid
250 mL
Chemical Substrate
Approved for shipment at Room Temperature or on Wet Ice
Colorimetric
Novex™
BCIP⁄NBT
AP (Alkaline Phosphatase) Substrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Novex AP Chromogenic Substrate (BCIP

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Maleic acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.