Learn More
Invitrogen™ Blotting Tweezer
For Bolt™ Mini Blot Module
Brand:  Invitrogen™ B1002
Product Code. 15341707
  • 26.90 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Gasket fits on the cathode core (-) of the blot module. Tweezers designed for handling nitrocellulose or PVDF membranes.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Protein assays and analysis, protein gel electrophoresis
Mini
1 unit
Bolt Mini Blot Module
Tweezers