missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Digitonin (5%)

Digitonin (5%), or digitin, is a non-ionic detergent with a cholesterol-like structure that can water-solubilize lipids, permeabilize cells, be used as a cytotoxicity positive control and solubilize membrane-bound proteins for NativePAGE™ protein analysis or cryo-EM structural analysis.

Brand:  Invitrogen™ BN2006

140.60 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.



Product Code. 10636033


Includes: 1 mL of solution
This item is not returnable. View return policy

Description

Description

Digitonin, also known as digitin, is a mild, non-ionic detergent with a cholesterol-like structure that allows it to water-solubilize lipids. It is useful for permeabilizing cells and can be used as a cytotoxicity chemistry positive control. In addition, it preserves the native lipid environment in membrane protein complexes and can be used to solubilize membrane-bound proteins. Digitonin has a molecular formula of C56H92O29 and molecular weight of 1229 Daltons.

Digitonin detergent solution has many more applications. It is used to prepare native protein samples for analysis on the NativePAGE Gel System. Included in the NativePAGE Sample Prep Kit (Cat No. BN2008), this ready-to-use detergent solution improves solubility of hydrophobic and membrane proteins during sample preparation. Samples prepared with 10% DDM, 5% digitonin, or the NativePAGE Sample Prep Kit are compatible with NativePAGE Novex Bis-Tris Gels for native electrophoresis and have increased resolution and reduced streaking. In addition, before structural determination by cryo-EM or crystallization, digitonin can be used to solubilize membrane proteins (typically at 1–2% w/v) and as a wash buffer.

It is important to note for sample preparation that the digitonin detergent micelle size is ∼70 kDa and is typically used at a concentration that is 1–2X the critical micelle concentration (CMC).

Furthermore, digitonin can be used as a spike in purified protein at CMC concentration to help with particle distribution and preferred orientation on grid during the vitrification process. In addition, digitonin provides efficient antibody penetration into tissue sections and is compatible with immunohistochemistry (IHC) staining. Digitonin at a final concentration of 0.2–0.5% in buffer is compatible with IHC.

NOTE: If the 5% digitonin solution precipitates out of solution, warm the solution at 37°C and gently shake to dissolve the precipitate.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

11024-24-1
Clear
IHC, Cryo-EM, Crystallography
Native-PAGE
1 mL
Native Gel Electrophoresis
1229
Digitonin
Store at 2–8°C.
5%
Mini Gel Tank, XCell SureLock Mini-Cell
Digitonin Solution
Liquid
NativePAGE Gels
Each
Wet Ice
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Digitonin (5%)

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P314-Get medical advice/attention if you feel unwell.
  • P330-Rinse mouth.

Supplemental information
  • MIXTURE LIST-Contains : digitonin

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.