missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gel-Dry™ Drying Solution
Modulates the rate of water release to ensure crack-free gels
46.67 € - 251.00 €
Specifications
Buffer | Drying Solutions |
---|---|
Content And Storage | Store at room temperature. |
For Use With (Equipment) | DryEase™ Mini-Gel Drying System |
Gel Size | Mini |
Product Line | Novex |
Product Code | Brand | Volume (Metric) Buffer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) Buffer | Price | Quantity & Availability | |||||
11599066
|
Invitrogen™
LC4025 |
500 mL |
46.67 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10482862
|
Invitrogen™
LC40254 |
8 x 500 mL |
251.00 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Gel-Dry™ Drying Solution is one component of the DryEase™ Mini-Gel Drying System which is a simple, easy-to-use kit for drying gels evenly using passive evaporation
- The kit allows gels to be assembled on a base between two open frames, so that both outer surfaces of the gels are exposed to air
- Consequently, drying is even and cracking is minimized
- The proprietary Gel-Dry™ Drying Solution modulates the rate of water release to ensure crack-free gels
- With the DryEase™ System, gels are dried overnight with uniform tension on all points of the gel to eliminate cracking
Applications
- Gels dried with the DryEase™ System are suitable for fluorography, densitometry, autoradiography, or permanent gel storage
Order Info
Shipping condition: Room Temperature
Specifications
Drying Solutions | |
DryEase™ Mini-Gel Drying System | |
Novex | |
Room Temperature |
Store at room temperature. | |
Mini | |
Drying Solution |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Gel-Dry Drying Solution
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : Ethanol, Methanol, Propan-2-ol
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title