missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HiMark™ Pre-stained Protein Standard

Designed for analysis of high molecular weight proteins on NuPAGE™ Novex™ Tris-Acetate gels

Brand:  Invitrogen™ LC5699


Product Code. 10237532

  • 393.00 € / 250µL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Includes: 250μL of HiMark™ Pre-Stained HMW Protein Standard; Storage Buffer: Tris-HCl, Formamide, SDS, Phenol Red
This item is not returnable. View return policy

Description

Description

  • Comprehensive - contains markers with a wide range of high molecular weights, providing nine protein bands in the range of 30 to 460kDa
  • Convenient - a ready-to-use formulation eliminates the need to heat, reduce, or add sample buffer prior to use
  • Efficient - allows easy visualization of electrophoresis and rapid evaluation of Western transfer efficiency
  • Compatible - intended for use with NuPAGE Novex 3 to 8% and 7% Tris-Acetate gels under denaturing conditions
  • Can also be used with NuPAGE Novex 4-12% Bis-Tris Gels with NuPAGE MOPS Running Buffer and Novex 4% Tris-Glycine Gel, however to obtain best results with HMW proteins, always use NuPAGE Novex Tris-Acetate Gels
  • Gel Compatibility: NovexTris-Glycine Gels, NuPAGE Tris-Acetate Gels

HiMark™ High Molecular Weight Protein Standard also available in an unstained format

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contents: 250 μL of HiMark™ Pre-Stained HMW Protein Standard
Storage Buffer: Tris-HCl, Formamide, SDS, Phenol Red
Storage:Store at -20°C
9
Yes
30 to 460 kDa
Novex™ Tris-Glycine Gels, NuPAGE™ Tris-Acetate Gels
HiMark™
Dry Ice
Colorimetric
Protein Ladder
Tris-acetate gels
2 colors: Blue, Pink
460, 268, 238, 171, 117, 71, 55, 41, 31 kDa
250 μL
Western Blotting, SDS-PAGE
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiMark Pre-stained Protein Standard

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : Formamide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.