Learn More
Invitrogen™ iBlot™ Transfer Stack, nitrocellulose, mini
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
iBlot Transfer Stacks are used to transfer proteins
224.00 € - 468.00 €
Specifications
Length (Metric) | 8 cm |
---|---|
Width (Metric) | 8 cm |
Format | Transfer Stack |
Material | Nitrocellulose |
Pore Size | 0.2 μm |
Includes: One box of iBlot™ Transfer Stacks containing 10 stacks.
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
10227972
![]() |
Invitrogen™
IB301002 |
Mini, 8 cm x 8 cm |
224.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
12219319
![]() |
Invitrogen™
IB301032 |
Mini, 8 cm x 8 cm |
468.00 €
Pack of 3 |
In Stock
Log in to see stock. |
|||||
Description
The iBlotTransfer Stacks are disposable stacks that come in three varieties for different applications: iBlotGel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins; iBlot DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlotTransfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.
Nitrocellulose membrane
- Composed of 100% pure nitrocellulose to provide high-quality transfer
- Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
- Bind proteins due to hydrophobic and electrostatic interactions
- Pore size: 0.2μm
- Protein binding capacity: 209μg/cm2
Transfer stack sizes
- The regular size (13 x 8.3cm) is used for all Novex midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels
- The mini size (8 x 8cm) is used for all Novex mini-gels (or equivalent)
Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room temperature
Specifications
8 cm | |
Transfer Stack | |
0.2 μm | |
iBlot |
8 cm | |
Nitrocellulose | |
Mini, 8 cm x 8 cm | |
Sustainable disposal |
Safety and Handling
- iBlot Transfer Stack, nitrocellulose, mini
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P314-Get medical advice/attention if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)-1,3-Propanediol ethylene glycol,aluminium oxide
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.