missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Large Gel Drying Frame

For use with Large Gel Drying Kit

Brand:  Invitrogen™ NI2200

Product Code. 10359182

  • 205.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket

Includes: One frame
This item is not returnable. View return policy

Description

Description

Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 frame; Store at room temperature
Novex™
Room Temperature
Large Gel Drying Frame
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.