Learn More
Invitrogen™ Large Gel Drying Kit
Accommodate larger format gels or four mini-gels
Brand:  Invitrogen™ NI2207
Product Code. 10573103
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: DryEase™ Large Gel Drying Frame (1 frame, 8 clamps), DryEase™ Large Pre-Cut Cellophane (100 sheets), and Gel-Dry™ Drying Solution (500mL).
Description
Description
The Large Gel Drying Kit dries gels using the same components as the DryEase™ Mini-Gel Drying System. Consistently dry gels without cracking and can accommodate up to four mini-gels or a large format gel. Base not required for assembly.
Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
The Large Gel Drying Kit contains a DryEase™ Large Gel Drying Frame (1 frame, 8 clamps), DryEase™ Large Pre-Cut Cellophane (100 sheets), and Gel-Dry™ Drying Solution (500 ml). Store solution at room temperature. Guaranteed stable for one year when properly stored.
Large Gel Drying Kit
Novex™
1 kit