missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NuPAGE™ LDS Sample Buffer (4X)
Used to prepare protein samples for denaturing gel electrophoresis with NuPAGE™ Novex™ gels. Invitrogen™ Novex™ NuPAGE™ LDS Sample Buffer (4X) contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.
Brand: Invitrogen™ NP0008
110.67 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Includes: NuPAGE LDS Sample Buffer (4X), 10mL
Description
Optimal protein sample preparation for polyacrylamide gel electrophoresis (PAGE) requires denaturing and reducing protein disulfide bonds.
- Highly viscous and concentrated solution that contains higher a concentration of glycerol
- Contains twice the amount of LDS compared to the amount of SDS in Novex™ Tris-Glycine SDS Sample Buffer or in Tricine SDS Sample Buffer
- NuPAGE™ LDS Sample Buffer should be brought to room temperature (25°C) prior to use
Order Info
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice
Specifications
Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red. Store at 4°C to 25°C. |
|
4X | |
LDS Sample Buffer | |
NuPAGE™ | |
Approved for shipment at Room Temperature or on Wet Ice |
Sample Loading Buffers | |
NuPAGE | |
NuPAGE™ Gels | |
250mL |
For Research Use Only. Not for use in diagnostic procedures.