missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ MES SDS Buffer Kit (for Bis-Tris Gels)

Product Code. 10683853
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10683853

Brand: Invitrogen™ NP0060

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

NuPAGE MES SDS Running Buffer (20X, 500mL), NuPAGE Sample Reducing Agent (10X, 250 μL) containing 500mM Dithiothreitol, NuPAGE Antioxidant (15mL), NuPAGE LDS Sample Buffer (10mL)

Requires

The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.

Designed for separation of small- to medium-size proteins on NuPAGE Novex Bis-Tris gels

Pre-mixed NuPAGE buffers are a convenient way to ensure high-quality, consistent electrophoresis results. All buffers are made with high-purity reagents and are strictly quality controlled. The buffers are provided as concentrated solution and require simple dilution with deionized water before use.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Product Type Buffer Kit
Content And Storage Contains
• NuPAGE™ MES SDS Running Buffer (20X, 500 ml)
• NuPAGE™ Sample Reducing Agent (10X, 250 μl) containing 500 mM Dithiothreitol
• NuPAGE™ Antioxidant (15 ml)
• NuPAGE™ LDS Sample Buffer (10 ml)
Store each at 4°C. Warm NuPAGE™ LDS Sample Buffer to 25°C before use.
Gel Type SDS PAGE Gel
Gel Compatibility Novex Tricine Gel
Product Line NuPAGE
Quantity 1 Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.