missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ MES SDS Running Buffer (20X)

Made with high-purity reagents and are strictly quality controlled

119.00 € - 878.00 €

Specifications

Chemical Name or Material Running Buffer
Concentration 20X
pH 7.3
Physical Form Liquid
Product Line NuPAGE
View More Specs

Includes: NuPAGE MES SDS Running Buffer (20X)

Requires: The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.


Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11509166
View accessory products View Documents
Invitrogen™
NP0002
500 mL
119.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11519166
View accessory products View Documents
Invitrogen™
NP000202
5 L
878.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NuPAGE MES SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Novex Bis-Tris gels only. NuPAGE MES SDS Running Buffer is recommended for separating small- to medium-sized proteins.

  • Use of MES buffer allows proteins to run faster
  • Ensures high-quality, consistent electrophoresis results

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Running Buffer
7.3
NuPAGE
20X
Liquid
1X: 50 mM MES, 50 mM Tris Base, 0.1% SDS, 1 mM EDTA, pH 7.3
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Contents: NuPAGE™ MES SDS Running Buffer (20X)
Storage: +4°C to 25°C

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.