Learn More
Invitrogen™ NuPAGE™ Sample Reducing Agent (10X)
Ready-to-use reagent to reduce protein samples for protein gel electrophoresis
20.31 € - 108.00 €
Specifications
Content And Storage | NuPAGE™ Sample Reducing Agent (500 mM dithiothreitol) should be stored at 4°C. Guaranteed stable for 6 months unless otherwise specified in the product literature. |
---|---|
Concentration | 500 mM |
Product Type | Sample Reducing Agent |
Product Line | NuPAGE™ |
Shipping Condition | Wet Ice |
Includes: NuPAGE Sample Reducing Agent (500mM dithiothreitol)
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10424012
|
Invitrogen™
NP0004 |
250μL |
20.31 €
250µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11569166
|
Invitrogen™
NP0009 |
10mL |
108.00 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NuPAGE Sample Reducing Agent contains 500mM dithiothreitol (DTT) at a ready-to-use, 10X concentration in a stabilized liquid form. Use NuPAGE Sample Reducing Agent when running proteins on NuPAGE Novex Bis-Tris gels to maintain protein samples in a reduced state during electrophoresis.
1D Gel Electrophoresis, 2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
NuPAGE™ Sample Reducing Agent (500 mM dithiothreitol) should be stored at 4°C. Guaranteed stable for 6 months unless otherwise specified in the product literature. | |
Sample Reducing Agent | |
Wet Ice |
500 mM | |
NuPAGE™ |
Safety and Handling
missing translation for 'shelfLife' : 6 Months
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.