Learn More
Invitrogen™ NuPAGE™ Transfer Buffer (20X)
Use to transfer proteins from NuPAGE Novex gels to membranes for Western blotting
Brand: Invitrogen™ NP0006
Description
NuPAGE Transfer Buffer can be used with the Novex Semi-Dry Blotter for semi-dry transfer or with the XCell II™ Blot Module for wet (tank) transfer. NuPAGE Antioxidant may be used with NuPAGE Transfer Buffer to enhance transfer of reduced proteins to membranes.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room Temperature

Specifications
Store at room temperature. Guaranteed stable for 6 months unless otherwise specified in the product literature. | |
20X | |
Transfer Buffer | |
NuPAGE™ | |
Room Temperature |
Transfer Buffers | |
NuPAGE | |
NuPAGE™ Gels | |
125mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.