missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ IEF Protein Gels, pH 3-10, 1.0 mm

Novex™ pH 3-10 IEF Gels are used for pI determination and are excellent for native applications using soluble proteins. Isoelectric focusing (IEF) is an electrophoresis technique that separates proteins based on their isoelectric point (pI).

Brand:  Invitrogen™ EC6655BOX

230.85 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 10741264

  • 243.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: One box of 5 gels
This item is not returnable. View return policy

Description

Description

Novex™ pH 3-10 IEF Gels are used for pI determination and are excellent for native applications using soluble proteins. Isoelectric focusing (IEF) is an electrophoresis technique that separates proteins based on their isoelectric point (pI). The pI is the pH at which a protein has no net charge and does not move in an electric field. Vertical IEF gels effectively create a pH gradient so proteins separate according to their unique pI.

IEG gels can be used to readily detect minor changes in a protein due to deamination, phosphorylation, or glycosylation, and can resolve different proteins of similar size that cannot be resolved on standard SDS-PAGE gels. Pre-focusing is not required. Total run time is approximately 2.5 hours.

Formulation
Novex™ IEF gels are 5% polyacrylamide and consist of high-purity acrylamide, bisacrylamide, TEMED, APS, ultrapure water, and 2% ampholytes. They do not contain denaturing reagents.

Recommended buffers
IEF is a sensitive technique which is affected by many factors. Our optimized, premixed IEF buffers reduce variability and help you obtain consistent results.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

XCell SureLock Mini-Cell
IEF
5 Gels/Box
1.0 mm
10-well
Store at 2°C to 8°C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number12120130 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberEC6655BOX
Lot Number12121231 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberEC6655BOX
Lot Number13041251 Certificate TypeCertificate of Analysis Date14/04/2017 Catalog NumberEC6655BOX
Lot Number12090731 Certificate TypeCertificate of Analysis Date13/04/2017 Catalog NumberEC6655BOX
Lot Number12051561 Certificate TypeCertificate of Analysis Date13/04/2017 Catalog NumberEC6655BOX
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Novex™ IEF Protein Gels, pH 3-10, 1.0 mm > 10-well

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.