missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tau (Total) Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ KMB7011

Product Code. 10736333

  • 722.00 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket

Includes:

Two vials Ms Tau (total) Standard; 25mL Standard Diluent Buffer; 1 plate (12 x 8 well strips), antibody-coated wells; 11mL Ms Tau (total) Detection Antibody; 0.125mL Anti-rabbit IgG HRP (100X); 25mL HRP Diluent; 100mL Wash Buffer Concentrate 25X); 25mL Stabilized Chromogen, Tetramethylbenzidine; 25mL Stop Solution; three Plate Covers

This item is not returnable. View return policy

Description

Description

The Mouse Tau (Total) ELISA quantitates Ms Tau in human brain homogenates, cell lysates, buffered solutions, or cell culture media. The assay will exclusively recognize both natural and recombinant MS Tau, independent of phosphorylation state. Principle of the method The Mouse Tau solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclindependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

P10637
<6.5 pg/mL
Biotin
Brain Homegenate, Cell Lysates, Supernatant
Colorimetric Microplate Reader
AI413597,AW045860,Mtapt,Tau
6%
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Tau
RUO
2°C to 8°C
1 hr. 20 min.
46.9-3000 pg/mL
31 to 2000pg/mL
ELISA Kit
Mouse
17762
4 hr.
5.8%
HRP
96 Tests
Brain Homogenate,2 μL; Cell Lysate,2 μL; Supernatant, 50 μL
Mouse
4 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.