missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SeeBlue™ Plus2 Pre-stained Protein Standard

Allows you to easily visualize protein molecular weight ranges during electrophoresis and quickly evaluate Western transfer efficiency

Brand:  Invitrogen™ LC5925

195.34 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.



Product Code. 10174502

  • 293.00 € / 500µL
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price


Includes: 500μL (50 applications of 10μL each) provided in a plastic vial Loading Buffer consists of Tris-HCl, Formamide, SDS, and Phenol Red
This item is not returnable. View return policy

Description

Description

The SeeBlue™ Plus2 Pre-Stained Standard contains 10 proteins: 8 blue-dyed and 2 with contrasting colors, providing easier band identification. It can be used with NuPAGE™, tris-glycine, and tricine gels.

  • 10 polypeptides resolved into 8 blue and 2 colored bands in the range of 4 to 250kDa (depending upon the buffer system)
  • Immediate, unambiguous band identification
  • A ready-to-use format—no mixing, heating, or reducing required
  • Gel Compatibility: Bolt Bis-Tris Plus Gels, NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels, NuPAGE Tris-Acetate Gels

This standard is pre-reduced and does not contain reducing reagents in the loading buffer.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 μL (50 applications of 10 μL each) provided in a plastic vial. Loading Buffer consists of Tris-HCl, Formamide, SDS, and Phenol Red.

Store at 4°C.
10
Yes
3 colors: Blue, Yellow, Pink
198, 98, 62, 49, 38, 28, 17, 14, 6, 3 kDa
500 μL
Western Blotting, SDS-PAGE
Colorimetric
Protein Ladder
3 to 200 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels
SeeBlue™
Wet Ice
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SEEBLUE PLUS 2 PRE-STAINED STANDARD

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : formamide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.