missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Sharp Unstained Protein Standard

Product Code. 10273723
Click to view available options
Quantity:
2 x 250 μL
Unit Size:
500µL
This item is not returnable. View return policy

Product Code. 10273723

Brand: Invitrogen™ LC5801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for accurate, easy and convenient molecular weight estimation of a wide range of molecular weight proteins during SDS-PAGE and western blotting

  • Accurate - Sharp bands for accurate estimation of molecular weigh
  • Comprehensive - A wide range of molecular weight bands consisting of 12 proteins in the range of 3.5 - 260kDa
  • Convenient - A ready-to-use formulation eliminates the need to heat, reduce, or add sample buffer prior to use
  • Adaptable - Suitable for most gel types, recommended for use with Novex™ NuPAGE™, Tris-Glycine, and Tricine Gels
  • Compatible with common gel stains - Works with SimplyBlue™, Coomassie™, silver and SYPRO™ staining method
  • Gel Compatibility: NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels, NuPAGE Tris-Acetate Gels, SDS-PAGE Gels

The Novex Sharp Protein Standard is also available in a pre-stained format.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contents: two vials of 250 μL each

Storage buffer: 65 mM Tris (pH 6.8), 30% glycerol, 2% SDS, 2.5 mM EDTA, 0.0025% Coomassie G-250

Storage: Upon receipt store at -20°C

Number of Markers 12
Product Type Protein Ladder
Ready to Load Yes
Size Range 3.5 to 260 kDa
Stain Type Unstained
Gel Compatibility Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 260, 160, 110, 80, 60, 50, 40, 30, 20, 15, 10, 3.5 kDa
Product Line Novex
Quantity 2 x 250 μL
Shipping Condition Dry Ice
System Type Western Blotting, SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.