missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNF alpha Rat ELISA Kit

Designed to detect and quantify the level of Rat TNF-α in serum, plasma, and supernatant using 96-well plates and a microplate reader

Brand:  Invitrogen™ KRC3011

 View more versions of this product

Product Code. 10640354

  • 668.00 € / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
This item is not returnable. View return policy

Description

Description

The Rat Tumor Necrosis Factor alpha (Rt TNFα) ELISA quantitates Rt TNFα in rat serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt TNFα. Principle of the method The Rat TNFα solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Tumor Necrosis Factor is a polypeptide cytokine produced by monocytes and macrophages. It functions as a multipotent modulator of immune response and further acts as a potent pyrogen. TNF-alpha circulates throughout the body responding to stimuli (infectious agents or tissue injury), activating neutrophils, altering the properties of vascular endothelial cells, regulating metabolic activities of other tissues, as well as exhibiting tumoricidal activity by inducing localized blood clotting. TNF-alpha also inhibits lipoprotein lipase activity resulting in cachexia, a physical wasting condition. Activation of B-cells by the Epstein Barr virus can be inhibited by TNF-alpha. Due to its varied actions throughout the immune system, TNF-alpha may play a role in the pathogenesis of many disease states. TNF-alpha production is mediated by the action of lymphokines and endotoxins on the macrophage. Purified monocytes produce TNF-alpha within four hours of stimulation by recombinant IL-2 and there is some in vitro evidence to suggest that TNF-alpha is expressed at high levels and with prolonged kinetics in T cells stimulated by both CD2 and CD28. Secretion of TNF-alpha is enhanced by gamma interferon. TNF then induces or enhances the specific production of Class I MHC antigen, GM-CSF, and IL-1.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

P16599
<4 pg/mL
ELISA Kit
Rat
Colorimetric Microplate Reader
RATTNF,TNF-alphaa,Tnf
5.8%
HRP
96 Tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Rat
4 hr.
11.7-750 pg/mL
Biotin
Plasma, Serum, Supernatant
Colorimetric
24835
8.3%
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
<4pg/mL
RUO
2°C to 8°C
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.