missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X)

Formulated for separation of proteins, in their denatured state, on Novex Tris-Glycine gels

Brand:  Invitrogen™ LC26755

175.68 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11569066

  • 288.00 € / 5 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: Novex Tris-Glycine SDS Running Buffer (10X), 500mL

Requires : The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.
This item is not returnable. View return policy

Description

Description

Tris-Glycine gels provide reproducible separation of a wide range of proteins into well-resolved bands, depending upon the gel percentage used.

  • Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results
  • Made with high-purity reagents and are strictly quality controlled

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Running Buffer
Novex Tris-Glycine Gels
Novex
10X
Liquid
5 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.