Learn More
Invitrogen™ XCell SureLock™ Mini-Cell and XCell II™ Blot Module
Quickly and easily run and blot Novex minigels
Brand: Invitrogen™ EI0002
1303.90 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: XCell II Blot Module; Gel Tension Wedge; Buffer Dam; Lower Buffer Chamber; Buffer Core; Mini-Cell Lid; Gel Knife
Description
The XCell SureLock Mini-Cell and XCell II Blot Module includes the XCell SureLock Mini-Cell for running Novex minigels and the XCell II Blot Module for blotting Novex minigels. The XCell SureLock Mini-Cell allows you to run Novex minigels quickly, easily, and leak-free without any clamps or grease. The incorporation of a gel tension wedge, in place of the rear wedge used on earlier models, makes the XCell SureLock Mini-Cell easy-to-use.
How the system works
- The XCell SureLock Mini-Cell holds gels firmly with a simple gel tension wedge
- When the lever on the gel tension wedge is pushed forward into the locked position, an even, horizontal force is generated, sealing the gel/buffer core assembly firmly into position in the lower buffer chamber
- The positive locking action of the gel tension wedge ensures a trouble-free, leak-free gel run every time
- The XCell SureLock Mini-Cell can be easily set up in just three simple steps
- The XCell SureLock Mini-Cell is available separately or as a part of a complete package
- Allows easy transfer of proteins or nucleic acids from minigels to membranes
- Fits neatly into the XCell SureLock and XCell II Mini-Cells in place of the gel/buffer core assembly and requires less than 200mL of transfer buffer for Western, Southern, and Northern transfers
- Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders
- Maximum blot size is 9 x 9cm
Upgrade XCell II to XCell SureLock Mini-Cell
- Conversion of an XCell II into an XCell SureLock is easy with the SureLock Retrofit Kit (available separately)
- Kit consists of a SureLock Mini-Cell lid, gel tension wedge, and molded minigel buffer dam
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room temperature

Specifications
Contains: • XCell II™ Blot Module • 1 Gel Tension Wedge • 1 Buffer Dam • 1 Lower Buffer Chamber • 1 Buffer Core • 1 Mini-Cell Lid • 1 Gel Knife Warrantied for one year from the time of purchasing. |
|
Wet Transfer | |
Mini Precast Protein Gels | |
Nitrocellulose, PVDF | |
Novex, XCell II, XCell SureLock | |
1 unit |
Mini Precast Protein Gels | |
XCell SureLock™ Mini-Cell | |
Mini (8 x 8 cm) | |
Wet | |
Blot Module | |
Vertical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.