missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZOOM™ Cathode Buffer 10X pH 9-12

Product Code. 11595076
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11595076

Brand: Invitrogen™ ZB10912

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

The ZOOM IEF Fractionator Combo Kit includes the ZOOM IEF Fractionator; one pack ZOOM Disks of each pH; and ZOOM Reagents (urea, thiourea, CHAPS, carrier ampholytes pH 3-10, anode buffers, and cathode buffers)

Use with ZOOM IPG Strips

IsoElectric Focusing, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cathode Buffer
Concentration 10 X
Content And Storage The ZOOM™ IEF Fractionator Combo Kit includes the ZOOM™ IEF Fractionator; one pack ZOOM™ Disks of each pH; and ZOOM™ Reagents (urea, thiourea, CHAPS, carrier ampholytes pH 3-10, anode buffers, and cathode buffers). ZOOM™ Disks, Carrier Ampholytes, and Buffers are stored at +4°C.
Gel Type IEF Gel
Shipping Condition Wet Ice
Gel Compatibility ZOOM IPG Strip
Product Line ZOOM
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.