Learn More
Novus Biologicals™ Inorganic Pyrophosphatase/PPA1 Protein
Highly purified. Generating reliable and reproducible results.
Brand:  Novus Biologicals™ NBP1-72378-100UG
Product Code. 18206801
  • 394.00 € / 100µg
Explore more special offers
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-176 of E.coli Inorganic Pyrophosphatase/PPA1 The Recombinant E. coli Inorganic Pyrophosphatase/PPA1 Protein is derived from E. coli. The Recombinant E. coli Inorganic Pyrophosphatase/PPA1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
SDS-PAGE
21.9kDa
100 μg
Store at -80°C. Avoid freeze-thaw cycles.
Inorganic Pyrophosphatase/PPA1
>95%
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 50mM NaCl
SDS-PAGE
Bacteria