missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NP-40 Surfact-Amps™ Detergent Solution

Código de producto. 11596671 Tienda Thermo Scientific Productos
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mL
500 mL
6 x 10 mL
missing translation for 'unitSize'
500mL
50mL
Pack of 6
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11596671

missing translation for 'mfr': Thermo Scientific™ 28324

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Thermo Scientific™ Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized and supplied as a 10% solution.

Thermo Scientific™ NP-40 Detergent Surfact-Amps Solution is highly-purified NP-40 detergent stabilized and supplied as a 10% solution.

This is an easy-to-use 10% (w/v) solution of purified NP-40 detergent for use in routine and high-demand protein research methods and molecular biology techniques. NP-40 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.

Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

NP-40 – a nonionic detergent for use in various protein methods

Accurate – precise 10% detergent solution in ultrapure water

Easy-to-use – solution is simple to dispense and dilute for use

Exceptionally pure – less than 1.0μeq/mL peroxides and carbonyls

Stable – packed in glass ampules under inert nitrogen gas

Detergent Properties:

  • Molecular Weight: 617
  • Detergent Class: Nonionic
  • Aggregation Number: 149
  • Micelle Molecular Weight: 90,000
  • Critical Micelle Concentration (CMC): 0.29mM (0.0179%, w/v)
  • Cloud Point: 80°C
  • Dialyzable: No

 

TRUSTED_SUSTAINABILITY

Especificaciones

Product Type Detergent
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Form Liquid
Product Line Surfact-Amps
Reagent Type Detergent Solution
Quantity 6 x 10 mL
Format Glass Ampule
compliance-icons
Nombre del producto
  • Surfact-Amps NP-40
Palabra de advertencia
  • Warning
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Indicaciones de peligro
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Otros peligros
  • MIXTURE LIST-Contains : Nonylphenol ethoxylates

For Research Use Only. Not for use in diagnostic procedures.

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado