Learn More
NQ301, MedChemExpress
MedChemExpress NQ301 is an antithrombotic agent; inhibits collagen-challenged rabbit platelet aggregation with an IC50 of 10 mg/mL.
Brand:  Medchem Express HY-101054
Encompass_Preferred
 View more versions of this product
Product Code. 16429796
  • 383.40 € / 50mg
Explore more special offers
Description
Description
NQ301 is an antithrombotic agent
Specifications
Specifications
NQ301
Red
C18H12ClNO3
DMSO : ≥ 29 mg/mL (89.03 mM)
325.75
98.12%
Solid
130089-98-4
Research
50 mg
O=C1C(NC2=CC=C(C(C)=O)C=C2)=C(Cl)C(C3=C1C=CC=C3)=O
325.75
Research
COVID-19-immunoregulation