Learn More
Abnova™ NRCAM 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand:  Abnova™ H00004897-T02.100uL
Product Code. 16138491
  • 215.00 € / 100µL
Explore more special offers
Specifications
293T
NRCAM 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 85.3kDa
Transient overexpression cell lysate was tested with anti-NRCAM antibody (Mfr. No. H00004897-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
BC098401.1
pCMV-NRCAM full-length
100 μL
Human
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abnova™ NRCAM 293T Cell Transient Overexpression Lysate (Denatured) > 100μL