missing translation for 'onlineSavingsMsg'
Learn More
NS 1738, Tocris Bioscience™ Product Code.: 18015572

NS 1738, Tocris Bioscience™

Product Code. 18015572
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18015572

Brand: Tocris Bioscience 2995/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulator of α7 nAChR; active in vivo

Selective positive allosteric modulator of α7 nicotinic acetylcholine receptors. Exhibits no substantial activity for α4β2, α3β3 and α1-containing receptors. Displays cognitive-enhancing properties in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501684-93-1
Molecular Formula C14H9Cl2F3N2O2
Molecular Weight (g/mol) 365.133
InChI Key OUDXRNQPVSMGDW-UHFFFAOYSA-N
Synonym n-5-chloro-2-hydroxyphenyl-n'-2-chloro-5-trifluoromethyl phenyl urea, 1-5-chloro-2-hydroxyphenyl-3-2-chloro-5-trifluoromethyl phenyl urea, d0d4lf, urea, n-5-chloro-2-hydroxyphenyl-n'-2-chloro-5-trifluoromethyl phenyl, 1-5-chloro-2-hydroxy-phenyl-3-2-chloro-5-trifluoromethyl-phenyl-urea, 3-5-chloro-2-hydroxyphenyl-1-2-chloro-5-trifluoromethyl phenyl urea, n-3-chloro-6-hydroxy-phenyl-n'-2-chloro-5-trifluoromethyl-phenyl urea
PubChem CID 310378
IUPAC Name 1-(5-chloro-2-hydroxyphenyl)-3-[2-chloro-5-(trifluoromethyl)phenyl]urea
SMILES C1=CC(=C(C=C1C(F)(F)F)NC(=O)NC2=C(C=CC(=C2)Cl)O)Cl

Specifications

Quantity 10 mg
Formula Weight 365.13
Percent Purity 98%
Chemical Name or Material NS 1738
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
NS 1738, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.