missing translation for 'onlineSavingsMsg'
Learn More

NS 309, Tocris Bioscience™

Product Code. 18045532
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive modulator of KCa2 and KCa3.1 channels

Positive modulator of small- and intermediate- conductance Ca2+-activated K+ channels (KCa2 and KCa3.1 channels); increases Ca2+ sensitivity. Displays no activity at BK channels.

Chemical Identifiers

CAS 18711-16-5
Molecular Formula C8H4Cl2N2O2
Molecular Weight (g/mol) 231.032
InChI Key CVOUSAVHMDXCKG-UHFFFAOYSA-N
Synonym 6,7-dichloro-1h-indole-2,3-dione 3-oxime, 6,7-dichloro-3-hydroxyimino indolin-2-one, z-6,7-dichloro-3-hydroxyimino indolin-2-one, 6,7-dichloro-3-hydroxyamino indol-2-one, 3-oxime-6,7-dichloro-1h-indole-2,3-dione, 6,7-dichloro-3-hydroxyamino-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1,3-dihydro-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1h-indol-2-one, ns309 hplc, ns hplc , solid
PubChem CID 11637204
IUPAC Name 6,7-dichloro-3-(hydroxyamino)indol-2-one
SMILES C1=CC(=C(C2=NC(=O)C(=C21)NO)Cl)Cl

Specifications

Quantity 50 mg
Formula Weight 231.04
Percent Purity >98%
Chemical Name or Material NS 309
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'