missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSC 74859, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
NSC 74859 selectively inhibits STAT3 DNA-binding activity in vitro (IC50 values are 86, 160, and >300 μM for STAT3-STAT3, STAT1-STAT3 and STAT1-STAT1 DNA-binding activity respectively). NSC 74859 blocks growth and induces apoptosis preferentially in tumor cells that contain persistently active STAT3 in vitro and arrests tumor growth in a murine xenograft model in vivo.
Specifications
Quantity | 50 mg |
Formula Weight | 365.36 |
Percent Purity | >98% |
Chemical Name or Material | NSC 74859 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'