missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSC16168, MedChemExpress
MedChemExpress NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.
598.05 € - 4449.60 €
Specifications
Chemical Name or Material | NSC16168 |
---|---|
CAS | 6837-93-0 |
Color | Light Nude |
Purity Grade Notes | Research |
Molecular Formula | C17H15NO9S3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16479676
|
Medchem Express
HY-100690 |
50 mg |
2988.90 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16454416
|
Medchem Express
HY-100690 |
100 mg |
4449.60 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16445857
|
Medchem Express
HY-100690 |
5 mg |
598.05 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16413128
|
Medchem Express
HY-100690 |
10 mg |
996.30 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.Specifications
NSC16168 | |
Light Nude | |
C17H15NO9S3 | |
NC1=C2C(C=C(S(=O)(O)=O)C=C2OS(C3=CC=C(C)C=C3)(=O)=O)=CC(S(O)(=O)=O)=C1 | |
473.5 | |
Research | |
Cancer-programmed cell death |
6837-93-0 | |
Research | |
DMSO : 31.25 mg/mL (66.00 mM; Need ultrasonic) | |
473.5 | |
98.0% | |
Powder |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only