Learn More
NSC16168, MedChemExpress
MedChemExpress NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.
Brand: Medchem Express HY-100690
This item is not returnable.
View return policy
Description
NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.Specifications
NSC16168 | |
Light Nude | |
C17H15NO9S3 | |
DMSO : 31.25 mg/mL (66.00 mM; Need ultrasonic) | |
473.5 | |
98.0% | |
Powder |
6837-93-0 | |
Research | |
10 mg | |
NC1=C2C(C=C(S(=O)(O)=O)C=C2OS(C3=CC=C(C)C=C3)(=O)=O)=CC(S(O)(=O)=O)=C1 | |
473.5 | |
Research | |
Cancer-programmed cell death |
Research purposes only