missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NTF3 Polyclonal Antibody
GREENER_CHOICE

Chicken Polyclonal Antibody

Brand:  Invitrogen™ PA19520

Product Code. 11558621

  • 373.00 € / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PA1-9520 detects NT3 in human and mouse samples. PA1-9520 has been successfully used in ELISA procedures. PA1-9520 immunogen is a synthetic peptide based on the human NT3 protein.

The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NTF3
Polyclonal
Unconjugated
Ntf3
AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3
Chicken
IgY fraction
RUO
18205, 4908
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
1 mg/mL
PBS with 0.02% sodium azide
P20181, P20783
Ntf3
Synthetic peptide based on the human NT3 protein.
500 μg
Primary
Human, Mouse
Antibody
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.