missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS Nuc-Off Nucleases and Nucleic Acid Removal Spray

Non-toxic spray designed to remove nucleases and DNA.

Brand:  MP BIOMEDICALS 112461450

 View more versions of this product

Product Code. 30214853

  • 204.00 € / 450mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Biomedicals Nuc-Off Nucleases and DNA Removal Spray is a ready-to-use solution for fast and cost-effective removal of RNase, DNase and DNA contamination.

This non-toxic spray-type solution can effectively remove nucleases and DNA from the surface of:

  • laboratory consumables
  • instruments
  • other utensils

creating a clean environment for RNA or DNA-related experiments.

It is strongly recommended to regularly clean your:

  • pipettors
  • bench surfaces
  • instrument exterior

Application Notes:

Nuc-Off Nucleases and DNA Removal Spray is intended for effective and safe elimination of Nucleases and DNA that may cause false positive or false negative experimental results.

Usage Statement:

Research Use Only (RUO)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Removal Agent
450 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.