Learn More
Nuclear Receptor Buffer F
Nuclear Receptor Buffer F
Brand:  Invitrogen PV4547
Product Code. 10116403
  • 153.00 € / Each
Explore more special offers
Description
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nuclear Receptor Buffer
25 mL
Liquid
Nuclear Receptor Buffer F may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).
Approved for shipment at Room Temperature or on Wet or Dry Ice