Learn More
Macherey-Nagel™ NucleoBond™ Finalizer
Concentrate and desalinate plasmid and cosmid DNA eluates obtained by anion-exchange chromatography. The NucleoBond™ Finalizer is a quick, efficient alternative to centrifugation and perfect for all types of downstream applications.
61.25 € - 91.45 €
Specifications
For Use With (Application) Applications: Quick concentration and desalination of plasmid and cosmid DNA eluates obtained by anion-exchange chromatography, Typical downstream applications: transfection, in-vitro transcription, automated or manual sequencing, cloning, hybridization, and PCR.
Product Type Desalting tool
Size 2-50kbp
Test Time 5min.
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12710941
View Documents
Macherey-Nagel™
740519.20
2 x 30, 2 x 1 mL
61.25 €
Pack of 20
11922442
View Documents
Macherey-Nagel™
740520.20
20 x 30, 20 x 1 mL
91.45 €
Pack of 20
Description
Description
Features
  • Desalting tools for speeding up the time-consuming precipitation step after anion-exchange plasmid purification
  • Eliminates centrifugation steps for precipitation – time saving from >1 h to only 5 minutes
  • Two sizes available, to be combined with Midi and Maxi preparations
  • Highly concentrated plasmid DNA
  • No loss of DNA pellets or incomplete solubilization of hardly visible precipitates
Applications
  • Quick concentration and desalination of plasmid and cosmid DNA eluates obtained by anion-exchange chromatography
  • Typical downstream applications: transfection, in-vitro transcription, automated or manual sequencing, cloning, hybridization, and PCR
Specifications
Specifications
Applications: Quick concentration and desalination of plasmid and cosmid DNA eluates obtained by anion-exchange chromatography, Typical downstream applications: transfection, in-vitro transcription, automated or manual sequencing, cloning, hybridization, and PCR.
Desalting tool
2-50kbp
5min.