missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Maxi Plus EF

Purify endotoxin-free plasmid DNA with high yield. The NucleoBond™ Xtra Maxi Plus EF Kit represents the new generation of anion exchanger allowing high yield, ultra-fast. The Xtra kit contains NucleoBond™ Finalizers to speed up DNA precipitation.

477.00 € - 2210.00 €

Specifications

Isolation Technology Silica Membrane Technology
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
12748422
View Documents
Macherey-Nagel™
740426.50
50
2210.00 €
50 preps
Please to purchase this item. Need a web account? Register with us today!
12816200
View Documents
Macherey-Nagel™
740426.10
10
477.00 €
2.80kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Features
  • Endotoxin-free
  • Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Optimized column design – Maxi prep in 50 min.
  • Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
  • Improved silica material – typical yields of 1,000μg (Maxi)
  • NucleoBond™Xtra Plus EF kits contain NucleoBond™Finalizers to speed up DNA precipitation
Applications
  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications

Specifications

Silica Membrane Technology
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : 2-propanol/ethanol (diluted < 20 %)/sodium hydroxide solution (diluted < 2 %), dodecyl sulfate, sodi

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.