missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra Midi EF
Purify endotoxin-free plasmid DNA with high yield. The NucleoBond™ Xtra Midi EF Kit represents the new generation of anion exchanger allowing high yield, ultra-fast.
186.00 € - 892.00 €
Specifications
Buffer | Yes |
---|---|
Format | Midi Gravity-flow Column |
Isolation Technology | Anion Exchange Chromatography Technology |
RNase | Yes |
Sample Volume | <200, <400μL |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
12773550
|
Macherey-Nagel™
740420.10 |
10 |
186.00 €
10 reactions |
In Stock
Log in to see stock. |
|||||
12798412
|
Macherey-Nagel™
740420.50 |
50 |
892.00 €
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Features- Endotoxin-free
- Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Optimized column design – Midi prep in 45 min.
- Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
- Improved silica material – typical yields of 400μg (Midi)
Applications
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Yes | |
Anion Exchange Chromatography Technology | |
<200, <400μL | |
400μg |
Midi Gravity-flow Column | |
Yes | |
<300kbp |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : 2-propanol/ethanol(diluted < 20 %)/sodium hydroxide solution(diluted < 2 %),dodecyl sulfate,sodium s