missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Midi Plus EF

Purify endotoxin-free plasmid DNA with high yield. The NucleoBond™ Xtra Midi Plus EF Kit represents the new generation of anion exchanger allowing high yield, ultra-fast. The Plus kit contains NucleoBond™ Finalizers to speed up DNA precipitation.

Brand:  Macherey-Nagel™ 740422.50


 View more versions of this product

Product Code. 12718422

  • 1020.00 € / 50 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features
  • Endotoxin-free
  • Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Optimized column design –  Midi prep in 45 min.
  • Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
  • Improved silica material – typical yields of 400μg (Midi)
  • NucleoBond™ Xtra EF kits contain NucleoBond™ Finalizers to speed up DNA precipitation
Applications
  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications

Specifications

Yes
Anion Exchange Chromatography Technology
50
Yes
<300kbp
Midi Gravity-flow Column
Nucleo Bond Xtra Midi Plus EF
Nucleo Bond Xtra Midi Plus EF
< 200, < 400μL
400μg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : 2-propanol/ethanol (diluted < 20 %)/sodium hydroxide solution (diluted < 2 %), dodecyl sulfate, sodi

SDS
Documents

Documents

Certificates
Special Offers

Special Offers