Learn More
Macherey-Nagel™ Nucleodur™ C18 Pyramid HPLC CC Guard Columns
Brand:  Macherey-Nagel™ 761854.30
 View more versions of this product
Product Code. 12488587
  • 228.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
14%
3 μm
110Å
3 mm
Acetonitrile/Water
Silica
Guard Column
HPLC
Yes
L1
1 to 9
8 mm
CC Guard Column
NUCLEODUR™ C18 Pyramid
Acetonitrile/Water
Octadecyl
LC/MS due to low bleeding characteristics
Spherical
Yes