Learn More
Macherey-Nagel™ Nucleodur™ HILIC HPLC Column
Column Format: Analytical Column; Carbon Load: 7%; Pore Size: 110 Å; Column Phase: HILIC
564.00 € - 815.00 €
Specifications
Column Format | Analytical Column |
---|---|
Particle Shape | Spherical |
pH | 2 to 8.5 |
Pore Size | 110 Å |
Surface Area | 340 m²/g |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
12728872
|
Macherey-Nagel™
760551.20 |
125 mm | 2 mm | 5 μm |
646.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12718862
|
Macherey-Nagel™
760528.20 |
150 mm | 2 mm | 1.8 μm |
815.00 €
Each |
In Stock
Log in to see stock. |
|||
12708862
|
Macherey-Nagel™
760526.20 |
100 mm | 2 mm | 1.8 μm |
815.00 €
Each |
In Stock
Log in to see stock. |
|||
12708872
|
Macherey-Nagel™
760550.20 |
250 mm | 2 mm | 5 μm |
754.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12728852
|
Macherey-Nagel™
760521.20 |
30 mm | 2 mm | 1.8 μm |
713.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12748872
|
Macherey-Nagel™
760552.20 |
50 mm | 2 mm | 5 μm |
564.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12717840
|
Macherey-Nagel™
760523.20 |
50 mm | 2 mm | 1.8 μm |
713.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12747840
|
Macherey-Nagel™
760525.20 |
75 mm | 2 mm | 1.8 μm |
713.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12738872
|
Macherey-Nagel™
760551.30 |
125 mm | 3 mm | 5 μm |
656.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12718872
|
Macherey-Nagel™
760550.30 |
250 mm | 3 mm | 5 μm |
749.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12738852
|
Macherey-Nagel™
760521.30 |
30 mm | 3 mm | 1.8 μm |
713.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12727840
|
Macherey-Nagel™
760523.30 |
50 mm | 3 mm | 1.8 μm |
713.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12675969
|
Macherey-Nagel™
760552.30 |
50 mm | 3 mm | 5 μm |
564.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12458467
|
Macherey-Nagel™
760551.46 |
125 mm | 4.6 mm | 5 μm |
723.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12695969
|
Macherey-Nagel™
760554.46 |
100 mm | 4.6 mm | 5 μm |
723.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
10016494
|
Macherey-Nagel™
760550.46 |
250 mm | 4.6 mm | 5 μm |
795.00 €
Each |
In Stock
Log in to see stock. |
|||
12478467
|
Macherey-Nagel™
760553.46 |
150 mm | 4.6 mm | 5 μm |
723.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12707840
|
Macherey-Nagel™
760521.46 |
30 mm | 4.6 mm | 1.8 μm |
779.00 €
Each |
In Stock
Log in to see stock. |
|||
12685969
|
Macherey-Nagel™
760552.46 |
50 mm | 4.6 mm | 5 μm |
615.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12448467
|
Macherey-Nagel™
760551.40 |
125 mm | 4 mm | 5 μm |
656.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
10596415
|
Macherey-Nagel™
760523.46 |
50 mm | 4.6 mm | 1.8 μm |
779.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12438467
|
Macherey-Nagel™
760550.40 |
250 mm | 4 mm | 5 μm |
749.00 €
Each |
In Stock
Log in to see stock. |
|||
12748852
|
Macherey-Nagel™
760521.40 |
30 mm | 4 mm | 1.8 μm |
713.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12737840
|
Macherey-Nagel™
760523.40 |
50 mm | 4 mm | 1.8 μm |
713.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12468467
|
Macherey-Nagel™
760552.40 |
50 mm | 4 mm | 5 μm |
564.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
Specifications
Analytical Column | |
2 to 8.5 | |
340 m²/g | |
Silica | |
HILIC | |
HILIC | |
EC Column |
Spherical | |
110 Å | |
60°C | |
7% | |
No | |
Stainless Steel |