Learn More
Macherey-Nagel™ Nucleogel™ HPLC Column
Brand:  Macherey-Nagel™ 720094.40
 View more versions of this product
Product Code. 10206511
  • 795.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
50 μeq/g
2 mM potassium hydrogen phthalate buffer pH 5.6
HPLC Column
HPLC, Separation of Inorganic Anions
10 μm