Learn More
Macherey-Nagel™ Nucleogel™ HPLC Column
Brand:  Macherey-Nagel™ 719475
 View more versions of this product
Product Code. 10388251
  • 1225.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Polymer-based
Purification of Peptides, Large Proteins and Oligonucleotides, HPLC
8 μm
HPLC Column
Polymer
0.1 M Na2SO4 + 0.2 % NaN3
Polymer-based
Quaternized PEI