missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoMag™ RNA Kit
Isolate RNA through a magnetic-bead based platform. Macherey Nagel™ NucleoMag™ RNA Kit enables isolation of RNA from tissue and cell samples.
380.00 € - 1345.00 €
Specifications
Format | Highly Reactive Superparamagnetic Beads |
---|---|
For Use With (Application) | Rapid manual and automated small-scale preparation of highly pure total RNA from tissue or cell samples |
Includes | NucleoMag™ B-Beads, buffers, TCEP, RNase-free rDNase |
Product Type | NucleoMag RNA |
RNase | Yes |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
12757790
|
Macherey-Nagel™
744350.4 |
4 x 96 |
1345.00 €
384 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12883931
|
Macherey-Nagel™
744350.1 |
96 |
380.00 €
96 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Features- One-tube procedure minimizes risk of cross-contamination
- Small elution volumes ≥ 50μL
- Suitable for manual and automated processing
- Recombinant Dnase included
- Reducing agent TCEP included (no β-mercaptoethanol necessary)
Applications
- Rapid manual and automated small-scale preparation of highly pure total RNA from tissue or cell samples
Specifications
Highly Reactive Superparamagnetic Beads | |
NucleoMag™ B-Beads, buffers, TCEP, RNase-free rDNase | |
Yes | |
<120 min./96 Reaction |
Rapid manual and automated small-scale preparation of highly pure total RNA from tissue or cell samples | |
NucleoMag RNA | |
>200 nt | |
<30μg |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 2
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanaten,ethanol/tris-(2-carboxyethyl)phosphine hydrochloride, TCEP(