Learn More
Macherey-Nagel™ NucleoSEQ™ kit for dye terminator removal
Prefilled single spin columns for dye terminator removal.
Brand:  Macherey-Nagel™ 740523.10
 View more versions of this product
Product Code. 12768462
  • 33.55 € / 10 reactions
Explore more special offers
Description
Description
  • Contains NucleoSEQ™ columns, collection tubes, and a user manual
  • Designed for fast and effective clean-up of nucleic acids
  • Using gel exclusion in a convenient spin column format allows reliable removal of smaller molecules from nucleic acids
  • Impurities, for example, salts, excess of labels, nucleotides, traces of organic solvents, primers are retained by the column while nucleic acids of interest are recovered with high yield
  • The columns are pre-filled with size exclusion matrix
  • Maximum sample volume to be loaded onto the column: 20 μL
  • Columns with dry gel matrix can be stored at room temperature for 12 months
  • Columns with hydrated gel matrix can be stored at 4°C for 14 days
Specifications
Specifications
10 Preps
5 min/prep (without hydration of matrix)
Dye terminator removal