Learn More
Macherey-Nagel™ Nucleosil™ 100-5 CN HPLC CC Guard Columns
Brand:  Macherey-Nagel™ 721604.40
Product Code. 12728122
  • 178.00 € / Pack of 3
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
5%
5 μm
100Å
4 mm
n-heptane
Silica
Guard Column
HPLC
Yes
L10
2 to 8
8 mm
HPLC CC Guard Column
NUCLEOSIL™ CN
n-heptane
Cyano
HPLC
Spherical
Hydrophobic, p-p