Learn More
Macherey-Nagel™ Nucleosil™ 300-5 Standard C18 HPLC CC Guard Columns
Brand:  Macherey-Nagel™ 721608.40
Product Code. 12768122
  • 178.00 € / Pack of 3
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
6.5%
5 μm
300Å
4 mm
Acetonitrile/Water
Silica
Guard Column
HPLC
Yes
Yes
2 to 8
8 mm
HPLC CC Guard Column
NUCLEOSIL™ Standard C18
Acetonitrile/Water
Octadecyl
HPLC
Spherical
Hydrophobic
L1