Learn More
Macherey-Nagel™ Nucleosil™ SB CC Guard Columns
Brand:  Macherey-Nagel™ 721885.40
Product Code. 12728142
  • 178.00 € / Pack of 3
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
10%
5 μm
100Å
HPLC CC Guard Column
NUCLEOSIL™ SB
Diammonium phosphate
Quaternary ammonium
HPLC
Spherical
Ion Exchange
2 to 8
8 mm
∽ 1 Meq/g
4 mm
Diammonium phosphate
Silica
Guard Column
HPLC
Yes
L14