missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ NucleoSpin™ 96 Blood QuickPure, 96-Well Kit

Ultra-fast isolation of highly concentrated DNA from blood in proven 96‐well plate format.

580.00 € - 5855.00 €

Specifications

Content And Storage 18°C to 25°C
Format 96-well Plate
For Use With (Application) Isolation of DNA
Target DNA
View More Specs

Products 3
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
12758522
View Documents
Macherey-Nagel Bioanalysis™
740667.2
192 Preps
580.00 €
192 reactions
Please to purchase this item. Need a web account? Register with us today!
12778522
View Documents
Macherey-Nagel Bioanalysis™
740667.4
384 Preps
1090.00 €
384 reactions
Please to purchase this item. Need a web account? Register with us today!
12768522
View Documents
Macherey-Nagel Bioanalysis™
740667.24
2304 Preps
5855.00 €
2304 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NucleoSpin™ Blood QuickPure binding plates are to be used in a centrifuge with deep buckets allowing processing of NucleoSpin™ 96 well plates stacked on 96 well blocks. The method is based on a specially treated silica membrane which speeds up the procedure by a combined washing/drying step. The number of washing and drying steps is reduced from 3 to 1. Therefore the hands-on time is minimised. Lysis is achieved by incubation of whole blood in a solution containing large amounts of chaotropic ions in the presence of proteinase K. Appropriate conditions for binding of DNA to the silica membrane of the NucleoSpin™ Blood QuickPure binding strips are created by addition of ethanol to the lysate. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer and can be used directly for PCR, Southern blotting or any kind of enzymatic reaction. The kit allows isolation of 4 μg to 6 μg of pure genomic DNA from 200 μL whole blood with an A260/280 ratio between 1.60 and 1.90 and a typical concentration of 40 to 60 ng/μL.Kit components: NucleoSpin™ Blood QuickPure binding plates, buffers, proteinase K, accessories.

  • Silica membrane technology
  • Manual processing
  • Yield: 4 to 6 μg
  • Fragment size: 300 bp to 30 kb
  • Sample material: human blood or other body fluid (200 μL/300 μL), 107 lymphocytes
  • Elution volume: 75 to 100 μL
  • Binding capacity: 60 μg
  • Preparation time: 30 min/prep
Specifications

Specifications

18°C to 25°C
96-well Plate
Isolation of DNA
DNA
Videos
Safety and Handling

Safety and Handling

Hazard Category
  • Skin sensitiser Category 1

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.